Learn More
Thermo Scientific™ Membrane Lactose Glucuronide Agar
Detect, enumerate and differentiate Escherichia coli and other coliforms by single membrane filtration with this ready-to-use medium.
Brand: Thermo Scientific™ PO1016A
28.30 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Detect, enumerate and differentiate Escherichia coli and other coliforms by single membrane filtration with ready-to-use Thermo Scientific™ Oxoid™ Membrane Lactose Glucuronide Agar (MLGA) The presence of lauryl sulphate and lactose with phenol red allow the selective isolation and differentiation of Escherichia coli and coliforms respectively.
Tests for Escherichia coli and coliforms provide a sensitive means for detection of fecal contamination of drinking water. They are regarded as one of the most important routine microbial examinations for assessing the effectiveness of water treatment and disinfection and for monitoring water quality in distribution1. Membrane Lactose Glucuronide Agar (MLGA) supports water testing utilizing single membrane filtration.
- Isolates coliforms effectively when used with single membrane filtration method.
- Selectively inhibits Gram-positive organisms, Lauryl sulphate selects for coliforms.
- Reduces confirmatory tests for E. coli due to the specificity of the reactions in the medium.
- Facilitates easy interpretation of the test results due to addition of chromogenic BCIG (5-bromo-4-chloro-3-indolyl-β-D-glucuronide), E. coli appear as green colonies while other coliform bacteria form yellow colonies.
- Ready-to-use convenience of prepared media.
Identification of Escherichia coli and coliforms is facilitated through two biochemical reactions within the medium: Lactose fermentation is detected by the dye phenol red which gives yellow colonies when acid is produced. The chromogenic substrate BCIG is cleaved by the enzyme glucuronidase and produces a blue chromophore which builds up in the bacterial cells. Coliforms are lactose-positive so colonies will be yellow; Escherichia coli is both lactose-positive and possesses glucuronidase so will appear as green colonies.1
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Red, clear | |
Membrane Lactose Glucuronide Agar | |
United Kingdom | |
19.5g ± 1.0g | |
10/Pk. |
2°C to 10°C | |
90mm Plate | |
Agar | |
Flow Wrapped - Single Layer |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4504391 | Certificate of Analysis | 21/04/2025 | PO1016A |
4501729 | Certificate of Analysis | 07/04/2025 | PO1016A |
4500341 | Certificate of Analysis | 31/03/2025 | PO1016A |
4499867 | Certificate of Analysis | 24/03/2025 | PO1016A |
4497548 | Certificate of Analysis | 17/03/2025 | PO1016A |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
81Hours
4Minutes
45Seconds
26New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.