missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 6.0 Product Code.: 15443479

Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 6.0

Product Code. 15443479
250 mL
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15443479

Brand: Thermo Scientific Chemicals J60763.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145224-94-8
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.248
MDL Number MFCD00283848
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
PubChem CID 16218417
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
SMILES C1COCCN1CCS(=O)(=O)O.O

Specifications

Concentration 1.0 M
pH 6.0
Physical Form Liquid
Quantity 250 mL
Solubility Information Miscible with water.
Chemical Name or Material MES buffer soln.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14L534 Certificate TypeCertificate of Analysis Date06/03/2025 Catalog NumberJ60763.AP,J60763.AK
Lot NumberZ27K514 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberJ60763.AP,J60763.AK
Lot NumberX23K517 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberJ60763.AP,J60763.AK
Lot NumberW06K535 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog NumberJ60763.AP,J60763.AK
Lot NumberU24K501 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberJ60763.AP,J60763.AK

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals MES, 1.0M buffer soln., pH 6.0 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.