missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-5-amino-1-benzothiophene-2-carboxylate, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | methyl 5-aminobenzo b thiophene-2-carboxylate, 5-aminothionaphthene-2-carboxylic acid methyl ester, methyl 5-aminobenzothiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 5-amino-, methyl ester, maybridge3_004413, methyl 5-aminobenzo b thiophene-carboxylate, methyl5-aminobenzo b thiophene-2-carboxylate, methyl 5-azanyl-1-benzothiophene-2-carboxylate, methyl-5-amino-benzo b thiophene-2-carboxylate, methyl 5-amino-1-benzo b thiophene-2-carboxylate |
Formula Weight | 207.25 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 5-amino-1-benzothiophene-2-carboxylate |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methyl-5-amino-1-benzothiophene-2-carboxylate, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction