missing translation for 'onlineSavingsMsg'
Learn More
Methyl piperidine-4-carboxylate, 98%, Thermo Scientific Chemicals Product Code.: 15498318

Methyl piperidine-4-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 15498318
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15498318

Brand: Thermo Scientific Alfa Aesar H61754.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl piperidine-4-carboxylate is a reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation. It is used as a reactant for synthesis of antitubercular agents, aminopyrazine inhibitors, orally available naphthyridine protein kinase D inhibitors and a scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl piperidine-4-carboxylate is a reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation. It is used as a reactant for synthesis of antitubercular agents, aminopyrazine inhibitors, orally available naphthyridine protein kinase D inhibitors and a scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2971-79-1
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00190578
InChI Key RZVWBASHHLFBJF-UHFFFAOYSA-N
Synonym methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4Show MoreShow Less
PubChem CID 424914
IUPAC Name methyl piperidine-4-carboxylate
SMILES COC(=O)C1CCNCC1

Specifications

Color Colorless to Yellow
Density 1.06 g/mL
Boiling Point 85°C to 90°C
Flash Point 89°C (192°F)
Refractive Index 1.465
Quantity 25 g
Beilstein 224128
Solubility Information Slightly soluble in water.
Formula Weight 143.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl piperidine-4-carboxylate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR23I035 Certificate TypeCertificate of Analysis Date06/06/2022 Catalog NumberH61754.14,H61754.22
Lot NumberY08H044 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH61754.14,H61754.22
Lot NumberS13F053 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61754.14,H61754.22
Lot NumberN22H075 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61754.14,H61754.22
Lot NumberP10G015 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61754.14,H61754.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl piperidine-4-carboxylate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.