Learn More
Micronova™ PolyMitt™ Cleaning Mitt Wipe
A range of cleaning mitts that offer the versatility of a high quality wipe with the convenience and security of a glove for simplified cleaning around hard to reach areas in the cleanroom. Available in a variety of fabrics for specialized cleaning.
Brand: Micronova™ MW-11
Description
The Micronova PolyMitt™ Cleaning Mitt Wipes come in one standard size, designed to fit comfortably over a gloved hand, making it easier to clean around carts, shelving, tanks, tubing and other hard to clean areas in the cleanroom.
- Durable cleaning mitts for simplified cleaning
- Versatility of a high quality wipe with the convenience and security of a glove
- One size: fits comfortably over a gloved hand
- Designed to clean hard to reach areas in the cleanroom
- Available in a range of fabrics for specialized cleaning:
- NovaPoly — 100% smooth knit polyester provides excellent disinfectant dispersal
- PolySorb — 100% textured polyester for removing residues and dispersing disinfectant
- MegaTex — 100% non-woven polyamide is absorbent with a mildly abrasive surface
- Microfiber — 80% polyester, 20% polyamide is highly absorbent and effective wet or dry
- Available with or without a urethane foam layer
- Compatible with strong disinfectants
- Autoclavable
- Reusable
Specifications
Mitt Wipes | |
30.5 cm | |
20.3 cm | |
Autoclavable | |
White | |
Reusable | |
With a waffle weave for increased absobency | |
1/Pk., 24/Cs. | |
Laundry instructions -- Item may be laundered and reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180 degree C. |
12 in. | |
8 in. | |
Textured Polyester | |
ISO 9001:2008 Facility | |
M-Zone MittWipes™ | |
30.5 x 20.3 cm (12 x 8 in.) | |
Cleaning Mitt for Cleanroom and Controlled Environment Surfaces | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
034434-000 | Certificate of Compliance | 16/09/2022 | MW-11 |
033163-000 | Certificate of Compliance | 03/08/2022 | MW-11 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.