Learn More
Elma™ Modular Baskets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
Brand:  Elma™ 3000006017
 View more versions of this product
Product Code. 17851337
  • £11.80 / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Specifications
Small Basket
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
12 mm
21 mm
4 Sections
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Elma™ Modular Baskets > Modular Baskets and Lids; Diameter (Metric): 21 mm; Height (Metric): 12 mm; Section: 4 Sections