Learn More
Invitrogen™ DQ™ Green BSA - Special Packaging
Hydrolysis of substrate yields brightly fluorescent product
Brand: Invitrogen™ D12050
392.16 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
DQ Green BSA is a fluorogenic substrate for proteases. A strong fluorescence quenching effect is observed when proteins are heavily labeled with BODIPY dyes. Upon hydrolysis of the DQ Green BSA to single, dye-labeled peptides by proteases, this quenching is relieved, producing brightly fluorescent products.
Biochemical Proteasome Assays, Cell Analysis, Cell Viability, Proliferation and Function, Drug Discovery and Development, Endocytosis, Exocytosis and Phagocytosis, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteasome Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation
Order Info
Shipping Condition: Room temperature

Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Solid | |
5 x 1 mg | |
Protein-Based Substrate |
Fluorescence | |
DQ™ | |
Protease Substrate | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
42Hours
23Minutes
4Seconds
58New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.