missing translation for 'onlineSavingsMsg'
Learn More
Morpholine, 99+%, extra pure, Thermo Scientific Chemicals Product Code.: 10397900

Morpholine, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10397900
1 L, Glass Bottle
Click to view available options
Quantity:
5 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10397900

Brand: Thermo Scientific Acros 158680010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Morpholine, 99+%, C4H9NO, CAS Number-110-91-8, 1-oxa-4-azacyclohexane, diethyleneimide oxide, diethylene imidoxide, diethylene oximide, diethylenimide oxide, drewamine, p-isoxazine, tetrahydro, tetrahydro-1,4-oxazine, tetrahydro-2h-1,4-oxazine, tetrahydro-p-oxazine, 1L, 99% min. (GC), 27, 5

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydroShow MoreShow Less
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -5.0°C
Density 0.9900g/mL
Boiling Point 126.0°C to 130.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4530 to 1.4550
Quantity 1 L
Beilstein 27, 5
Fieser 01,705; 11,352; 13,14
Merck Index 15, 6362
Specific Gravity 0.99
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetone, benzene,, ether, castor oil, ethylene glycol, 2-hexanone,, linseed oil, turpentine, pine oil
Viscosity 2.23 mPa.s (20°C)
Formula Weight 87.12
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Morpholine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442866 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number158680010,158680100,158680050,158680025
Lot NumberA0463195 Certificate TypeCertificate of Analysis Date04/06/2024 Catalog Number158680010,158680100,158680050,158680025
Lot NumberA0435048 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number158680010,158680100,158680050,158680025
Lot NumberA0460121 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number158680010,158680100,158680050,158680025
Lot NumberA0458461 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number158680010,158680100,158680050,158680025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Morpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Morpholine, 99+%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.