missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Protease, from S. aureus strain V8
Used for selective cleavage of proteins for amino acid sequence determination or peptide mapping
Brand: MP Biomedicals™ 0215197205
This item is not returnable.
View return policy
Description
- Unit Definition: One unit will hydrolyze casein to change A280 0.001 per minute, pH 7.8, at 37°C.
- Extinction Coefficient: (E1%) E1%280 = 4.26
- Solubility: Soluble in aqueous buffers (such as 10 mM Tris, pH 7.8 at 1 mg/mL).
- CAS Number: 66676-43-5
- Molecular Weight: 27,700 (sedimentation equilibrium)

Specifications
Used for selective cleavage of Proteins for Amino Acid sequence determination or Peptide mapping | |
Staphylococcus aureus V8 Strain | |
Powder | |
Protease |
4 and 7.8 | |
-20°C | |
5 mg |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Protease
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Protease
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Protease, from S. aureus strain V8 > 5mg
Spot an opportunity for improvement?Share a Content Correction