missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Measures 150 x 150mm
Specifications
Sub-Gel Midi-Plus Horizontal Gel Electrophoresis | |
UV Tray | |
150 x 150 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Videos
-
Fisherbrand and Fisher BioReagents Focus on electrophoresis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ UV Tray >
Spot an opportunity for improvement?Share a Content Correction