Learn More
Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories
Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
Brand: Thermo Scientific™ 24072677
856.15 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.
Ordering Information:
- Cat. No. 24070297 and 24070290 include tip combs

Specifications
Tube Dispensing Cassette | |
Long standard tube dispensing cassette (tubing length 200cm) | |
5 to 2500 μL | |
<7 mL |
Autoclavable (50 times at 121°C, 1 bar, 20 min.) | |
Polypropylene Tip, Silicone Tubing | |
Multidrop Combi, Multidrop 384, Multidrop DW | |
Tip: 0.5 mm; Tubing: 1.3 mm |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
4Hours
7Minutes
52Seconds
29 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
21Hours
7Minutes
52Seconds
29
Your input is important to us. Please complete this form to provide feedback related to the content on this product.