missing translation for 'onlineSavingsMsg'
Learn More
N-(2-Bromoethyl)phthalimide, 97+%, Thermo Scientific Chemicals Product Code.: 11454793

N-(2-Bromoethyl)phthalimide, 97+%, Thermo Scientific Chemicals

Product Code. 11454793
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11454793

Brand: Thermo Scientific Alfa Aesar A12164.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.

Solubility
Soluble in water (224 mg/L at 25°C).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 574-98-1
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.083
MDL Number MFCD00005902
InChI Key CHZXTOCAICMPQR-UHFFFAOYSA-N
Synonym n-2-bromoethyl phthalimide, 2-2-bromoethyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-bromoethyl, 2-2-bromoethyl-1h-isoindole-1,3 2h-dione, beta-bromoethylphthalimide, 2-bromoethyl phthalimide, 1-bromo-2-phthalimidoethane, beta-phthalimidoethyl bromide, n-2-bromoethyl-phthalimide, 2-2-bromo-ethyl-isoindole-1,3-dioneShow MoreShow Less
PubChem CID 11325
IUPAC Name 2-(2-bromoethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCBr

Specifications

Melting Point 80°C to 84°C
Quantity 100 g
Beilstein 148736
Solubility Information Soluble in water (224mg/L at 25°C).
Formula Weight 254.09
Percent Purity ≥98%
Chemical Name or Material N- (2-Bromoethyl)phthalimide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244152 Certificate TypeCertificate of Analysis Date21/10/2022 Catalog NumberA12164.22,A12164.14,A12164.36
Lot Number10203349 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12164.22,A12164.14,A12164.36
Lot Number10149977 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12164.22,A12164.14,A12164.36
Lot Number10121407 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12164.22,A12164.14,A12164.36
Lot Number10137893 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12164.22,A12164.14,A12164.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(2-Bromoethyl)phthalimide, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.