missing translation for 'onlineSavingsMsg'
Learn More
N-Methyl-N-[(1-methyl-1H-indol-6-yl)methyl]amine, 97%, Thermo Scientific™ Product Code.: 10317404

N-Methyl-N-[(1-methyl-1H-indol-6-yl)methyl]amine, 97%, Thermo Scientific™

Product Code. 10317404
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10317404

Brand: Thermo Scientific Maybridge CC44246DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-20-4
Molecular Formula C11H14N2
Molecular Weight (g/mol) 174.247
MDL Number MFCD08690256
InChI Key ODIDITAYVOGYIV-UHFFFAOYSA-N
Synonym n-methyl-n-1-methyl-1h-indol-6-yl methyl amine, methyl 1-methylindol-6-yl methyl amine, 1h-indole-6-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-6-yl methanamine, 1h-indole-6-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-6-ylmethyl-amineShow MoreShow Less
PubChem CID 18525770
IUPAC Name N-methyl-1-(1-methylindol-6-yl)methanamine
SMILES CNCC1=CC2=C(C=C1)C=CN2C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 174.25
Percent Purity 95%
Chemical Name or Material N-methyl-N-[(1-methyl-1H-indol-6-yl)methyl]amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methyl-N-[(1-methyl-1H-indol-6-yl)methyl]amine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.