missing translation for 'onlineSavingsMsg'
Learn More
(N,N-Dimethyl)methyleneammonium iodide, 97%, Thermo Scientific Chemicals Product Code.: 11492118

(N,N-Dimethyl)methyleneammonium iodide, 97%, Thermo Scientific Chemicals

Product Code. 11492118
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11492118

Brand: Thermo Scientific Alfa Aesar A15146.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(N,N-Dimethyl)methyleneammonium iodide is a useful reagent for many synthetic applications like aminomethylation. It is also essential for the conversion of ketones to α, β-unsaturated enones. It is utilized to prepare derivatives of the type RCH2N(CH3)2. It is extensively used in medicines like painkillers and sedatives- Tramadol hydrochloride intermediate viz. 2-dimethylamine methylcyclohexanone. It is also employed in reactive dyes, synthesized spices. It plays an essential role in Mannich reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(N,N-Dimethyl)methyleneammonium iodide is a useful reagent for many synthetic applications like aminomethylation. It is also essential for the conversion of ketones to α, β-unsaturated enones. It is utilized to prepare derivatives of the type RCH2N(CH3)2. It is extensively used in medicines like painkillers and sedatives- Tramadol hydrochloride intermediate viz. 2-dimethylamine methylcyclohexanone. It is also employed in reactive dyes, synthesized spices. It plays an essential role in Mannich reactions.

Solubility
Soluble in dimethylformamide. Partially soluble in acetonitrile, tetrahydrofuran and dichloromethane.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33797-51-2
Molecular Formula C3H8IN
Molecular Weight (g/mol) 185.008
MDL Number MFCD00011810
InChI Key VVDUZZGYBOWDSQ-UHFFFAOYSA-M
Synonym eschenmoser's salt, n,n-dimethylmethyleneiminium iodide, n-methyl-n-methylenemethanaminium iodide, dimethyl methylidene azanium iodide, n,n-dimethyl methyleneammonium iodide, dimethyl methylene ammonium iodide, methanaminium, n-methyl-n-methylene-, iodide, eschenmoser inverted exclamation mark s salt, eschenmoser salt, dimethyl methylene ammounium iodideShow MoreShow Less
PubChem CID 2724133
IUPAC Name dimethyl(methylidene)azanium;iodide
SMILES C[N+](=C)C.[I-]

Specifications

Melting Point ∼240°C (decomposition)
Quantity 50 g
Beilstein 3594966
Sensitivity Light and moisture sensitive
Merck Index 14,3251
Solubility Information Soluble in dimethylformamide. Partially soluble in acetonitrile,tetrahydrofuran and dichloromethane.
Formula Weight 185.01
Percent Purity 97%
Chemical Name or Material (N,N-Dimethyl)methyleneammonium iodide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250933 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15146.18,A15146.30,A15146.09
Lot Number10242691 Certificate TypeCertificate of Analysis Date12/08/2022 Catalog NumberA15146.18,A15146.30,A15146.09
Lot Number10159749 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15146.18,A15146.30,A15146.09
Lot Number10103005 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15146.18,A15146.30,A15146.09
Lot Number10159978 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15146.18,A15146.30,A15146.09
compliance-icons
Product Identifier
  • (N,N-Dimethyl)methyleneammonium iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : N,N-Dimethylmethyleneammonium iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(N,N-Dimethyl)methyleneammonium iodide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.