missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ LDPE, Round Carboy Product Code.: 10250831
Add to basket

Thermo Scientific™ Nalgene™ LDPE, Round Carboy

Product Code. 10250831
20 L
Click to view available options
Capacity (Metric):
10 L
15 L
20 L
25 L
50 L

Product Code. 10250831

Brand: Thermo Scientific™ 22100050

Edge
Add to basket
Add to basket

For Use With: Store and transport reagents; Closure Material: Polypropylene; Color: White

Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.

  • Molded-in graduations of 5 L and 1 gal. increments
  • Leakproof†

Includes:White polypropylene screw cap and 83B closure

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 83B
Color White
Description Round LDPE Carboy with Handle
Diameter (Metric) Inner Neck 64 mm
Diameter (Metric) Outer 286 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Height (Metric) With Closure 528 mm
Includes Handles
Material LDPE
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Handle
Shape Round
Sterility Non-Sterile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1424503 Certificate TypeCertificate of Compliance Date14/03/2025 Catalog Number2210-0050
Lot Number1293343 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2210-0050
Lot Number1287272 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2210-0050
Lot Number1409584 Certificate TypeCertificate of Compliance Date09/08/2024 Catalog Number2210-0050
Lot Number1402232 Certificate TypeCertificate of Compliance Date01/03/2024 Catalog Number2210-0050
  • Multiple Manufacturers
    Save up to 94% on Teaching Lab Essentials
    Promo Code25721

    Expires2025-05-01

    New Offer
    Save Now
  • Multiple Manufacturers
    Use code 24095 to receive this promotion.
    Promo Code: 24095

    Expires2025-04-15

    Days

    4

    Hours

    6

    Minutes

    24

    Seconds

    33
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 17/03/2025 and 15/04/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.

    Terms & Conditions Learn More

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ LDPE, Round Carboy >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.