Learn More
Whirl-Pak™ Standard Sample Bags
Features puncture proof tabs for protection from damage due to wire-end protrusion. Easy-to-close bags are ideal for sampling and product analysis. Available in plain or write-on styles.
£11.31 - £1419.35
Specifications
Autoclavable | Not Autoclavable |
---|---|
Disposable | Single-use |
Material | Polyethylene |
Sterility | Sterile |
No. per Pack | 500/Pk. |
Product Code | Brand | Product Type | Capacity (English) | Thickness (English) | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Capacity (English) | Thickness (English) | Dimensions (L x W) | Price | Quantity & Availability | |||||
11771664
![]() |
Whirl-Pak™
B01009 |
Sample Bag | 2 oz. | 2.25 mil | 12.5 x 7.5 cm |
£72.14
Pack of 500 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11701973
![]() |
Whirl-Pak™
B00679WA |
Sample Bag | 4 oz. | 2.25 mil | 18.5 x 7.5 cm |
£84.85
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11781664
![]() |
Whirl-Pak™
B00992 |
Sample Bag | 7 oz. | 3 mil | 18 x 9.5 cm |
£91.90
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11791664
![]() |
Whirl-Pak™
B01018 |
Sample Bag | 13 oz. | 3 mil | 19 x 13 cm |
£416.34
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11785714
![]() |
Whirl-Pak™
B00736WA |
Sample Bag | 18 oz. | 2.5 mil | 23 x 11.5 cm |
£124.80
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||||
11775774
![]() |
Whirl-Pak™
B01020WA |
Sample Bag | 24 oz. | 3 mil | 23 x 15 cm |
£134.00
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11701674
![]() |
Whirl-Pak™
B01063WA |
Sample Bag | 24 oz. | 4 mil | 23 x 15 cm |
£223.60
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11741684
![]() |
Whirl-Pak™
B00990 |
Sample Bag | 27 oz. | 3 mil | 30.5 x 12.5 cm |
£141.00
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11786508
![]() |
Whirl-Pak™
B00994WA |
Sample Bag | 36 oz. | 3 mil | 38 x 12.5 cm |
£160.00
Pack of 500 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
11791963
![]() |
Whirl-Pak™
B01027WA |
Sample Bag | 42 oz. | 3 mil | 38 x 15 cm |
£193.00
Pack of 500 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
|||||
11707093
![]() |
Whirl-Pak™
B01323WA |
Sample Bag | 69 oz. | 3 mil | 38 x 19 cm |
£221.00
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11776965
![]() |
Whirl-Pak™
B01067WA |
Sample Bag | 1 oz. | 2.25 mil | 12.5 x 6.5 cm | N/A | ||||||
11781963
![]() |
Whirl-Pak™
B01064WA |
Sample Bag with Labeling Area | 2 oz. | 2.25 mil | 12.5 x 7.5 cm |
£74.75
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11735804
![]() |
Whirl-Pak™
B01062WA |
Sample Bag with Labeling Area | 4 oz. | 2.25 mil | 18.5 x 7.5cm |
£90.90
Pack of 500 |
Estimated Shipment: 02-05-2025
Log in to see stock. |
|||||
11771963
![]() |
Whirl-Pak™
B01065WA |
Sample Bag with Labeling Area | 18 oz. | 2.5 mil | 23 x 11.5cm |
£315.68
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11761684
![]() |
Whirl-Pak™
B01196 |
Sample Bag with Labeling Area | 24 oz. | 4 mil | 23 x 15 cm |
£218.00
Pack of 500 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
11771684
![]() |
Whirl-Pak™
B01297WA |
Sample Bag with Labeling Area | 24 oz. | 3 mil | 23 x 15cm |
£150.94
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11734834
![]() |
Whirl-Pak™
B01491WA |
Sample Bag with Labeling Area | 27 oz. | 3 mil | 30.5 x 12.5 cm |
£169.00
Pack of 500 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11751684
![]() |
Whirl-Pak™
B01195 |
Sample Bag with Labeling Area | 55 oz. | 4 mil | 30 x 19cm |
£857.00
Pack of 500 |
In Stock
Log in to see stock. |
|||||
30286575
|
Whirl-Pak™
B01339WA |
Sample Bag with Labeling Area | 4 oz. | 2.25 mil | 18.5 x 7.5 cm |
£1419.35
Pack of 10000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Features puncture proof tabs for protection from damage due to wire-end protrusion. Easy-to-close bags are ideal for sampling and product analysis. Available in plain or write-on styles.
- Transparent polyethylene bags
- Whirling or folding thebag over three times and securing with the closure tabs creates a leak-proof sampling container
- Gas sterilized, with documentation available
- Pull tabs help prevent contamination from finger contact when opening
- Write-on bags have area that accepts ballpoint pen or pencil
- Bags can be frozen to any temperature; including liquid nitrogen--careful handling required after freezing
- Thickness, 2.25 to 4 mil
- RNase, DNase, pryogen and BPH free
- Comply with regulatory and industrial requirements of EPA, FDA, HACCP and USDA
- Bags should not be used at temperatures above 180°F (82°C)
Specifications
Not Autoclavable | |
Polyethylene | |
500/Pk. | |
Plastic | |
Ethylene Oxide Gas | |
-196°C to +82°C |
Single-use | |
Sterile | |
Cardboard | |
500/Pk. | |
-320°F to +180°F |