Learn More
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Brand:  Thermo Scientific™ C4013-195
 View more versions of this product
Product Code. 11985673
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes: 
100 vials with pre-attached caps and septa
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
Clear Glass, flanged; PP Black Cap; Red PTFE/White Silicone Preslit septum; in storage tray
Glass, PTFE, Silicone
2 mL
Storage Tray
Polypropylene, Flanged
Red/White
Clear
Yes
PTFE/Silicone, Pre-slit
Polypropylene
8-425
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits > Clear Glass, flanged; PP Black Cap; Red PTFE/White Silicone Preslit septum; in storage tray