missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nitrocellulose/Filter Paper Sandwich, 0.45 μm, 8.3 x 7.3 cm

Ideal for blotting proteins and nucleic acids

Brand:  Invitrogen™ LC2001

187.15 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 10493242

  • £204.00 / 5g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The 0.45μm pore size is ideal for the transfer of low molecular weight proteins (<20kDa) and nucleic acids (<300bp).

 

  • Composed of 100% pure nitrocellulose to provide high-quality transfer
  • Contain no support fabrics or detergents
  • Are compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, fluorescence, and radiolabeling
  • Provide high-sensitivity with low background
  • Are supplied in a pre-cut, pre-assembled membrane/filter paper sandwich for convenience
  • A protein's properties (i.e., charge, hydrophobicity, etc.) affect its ability to bind to membrane surfaces

 

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Proteins, Expression, Isolation and Analysis, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8.3 cm
Nitrocellulose Pre-Cut Blotting Membrane, 0.45μm pore size
Sandwich
0.45 μm
Room Temperature
0.8 mm
7.3 cm
20 membrane/filter paper sandwiches
Nitrocellulose
Store at room temperature.
7.3 cm x 8.3 cm
Novex
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2982998 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberLC2001
Lot Number3041825 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberLC2001
Lot Number2472023 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberLC2001
Lot Number2536358 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberLC2001
Lot Number2563756 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberLC2001
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Nitrocellulose/Filter Paper Sandwich, 0.45 μm, 8.3 x 7.3 cm > 0.45μm pore size; 20/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.