Learn More
Nucleoside-Analog-2, MedChemExpress
MedChemExpress Nucleoside-Analog-2 is a 4'-Azidocytidine analogue against Hepatitis C virus (HCV) replication.
Brand: Medchem Express HY-77652
Description
Nucleoside-Analog-2 is a 4'-Azidocytidine analogue against Hepatitis C virus (HCV) replication.Specifications
Nucleoside-Analog-2 | |
Gray | |
C9H11N5O6 | |
DMSO : 33.33 mg/mL (116.86 mM; Need ultrasonic) ∣H2O : 20 mg/mL (70.12 mM; Need ultrasonic) | |
285.21 | |
95.0% | |
Solid |
876708-01-9 | |
Research | |
50 mg | |
O[C@@H]1[C@](CO)(N=[N+]=[N-])O[C@@H](N2C(NC(C=C2)=O)=O)[C@H]1O | |
285.21 | |
Research | |
COVID-19-anti-virus |
Product Suggestions
Customers who viewed this item also viewed