Learn More
Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates

Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates.
Brand: Thermo Scientific™ 262146
151.80 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates. The clear and U shaped wells are ideal for solution-based assays (e.g. agglutination assays) and common serial dilutions, allowing clean and easy pipetting. Available in a variety of options for surfaces, sterility, closures, and packaging sizes.
- Clear polystyrene has excellent optical properties ideal for optical measurements
- Thermo Scientific™ Nunclon™ Delta is a hydrophilic surface treatment whereas the non-treated polystyrene is more hydrophobic
- Customized bar coding is available
Warranty: 90 days

Specifications
American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Without Lid | |
135 Pk. | |
Clear | |
Polystyrene | |
Non-sterile | |
Round Bottom (U-well) | |
135 |
Standard | |
8 x 12 | |
Nunc | |
Microplate | |
96 | |
Non-treated | |
135 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
179907 | Certificate of Compliance | 03/12/2024 | 262146 |
182187 | Certificate of Compliance | 03/12/2024 | 262146 |
178260 | Certificate of Compliance | 03/12/2024 | 262146 |
167674 | Certificate of Compliance | 03/12/2024 | 262146 |
172458 | Certificate of Compliance | 03/12/2024 | 262146 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.