Learn More
Thermo Scientific™ Nunc™ Microplate Lids



Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Brand: Thermo Scientific™ 263339
47.47 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
- Lids are for use with any standard plates
- 96 Well Lids have individual condensation rings
- Lids have notches for quick orientation
- Universal Lids can be used on any standard height plate on the market

Specifications
Clear | |
Polystyrene | |
96 | |
5 in. | |
Yes | |
100 | |
5 x 3.35 in. (127 x 85 mm) | |
3.35 in. | |
Plate Lid | |
None | |
All well shapes | |
Long Side |
5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm) | |
Non-sterile | |
7.9 mm | |
127 mm | |
0.3 in. | |
5 | |
Yes | |
85 mm | |
Nunc | |
-20°C to +80°C | |
96-well Plate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185146 | Certificate of Compliance | 19/03/2025 | 263339 |
185057 | Certificate of Compliance | 18/12/2024 | 263339 |
184438 | Certificate of Compliance | 06/12/2024 | 263339 |
176359 | Certificate of Compliance | 03/12/2024 | 263339 |
164375 | Certificate of Compliance | 03/12/2024 | 263339 |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.