missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Omeprazole, 98+% Product Code.: 15532343

Thermo Scientific Chemicals Omeprazole, 98+%

Product Code. 15532343
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15532343

Brand: Thermo Scientific Chemicals 458520250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73590-58-6
Molecular Formula C17H19N3O3S
Molecular Weight (g/mol) 345.42
InChI Key SUBDBMMJDZJVOS-UHFFFAOYSA-N
Synonym omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
PubChem CID 4594
ChEBI CHEBI:77260
IUPAC Name 6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC

Specifications

Melting Point 158.0°C to 159.0°C
Loss on Drying 0.5% max. (60°C, 4 h) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: very soluble in chloroform, dimethylformamide, slightly soluble in methanol,ethanol, very slighly soluble in acetonitrile, isopropanol
Formula Weight 345.42
Percent Purity 98+%
Chemical Name or Material Omeprazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451648 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number458520050,458520250,458520010
Lot NumberA0437647 Certificate TypeCertificate of Analysis Date12/01/2022 Catalog Number458520050,458520250,458520010
Lot NumberA0419428 Certificate TypeCertificate of Analysis Date29/05/2020 Catalog Number458520050,458520250,458520010
Lot NumberA0389501 Certificate TypeCertificate of Analysis Date05/05/2020 Catalog Number458520050,458520250,458520010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Omeprazole, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.