Learn More
Invitrogen™ OneComp eBeads™ Compensation Beads
Spherical particles that can be stained with individual fluorochrome-conjugated antibodies for use as single-color compensation controls. OneComp eBeads react with antibodies of mouse, rat and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test
Brand: Invitrogen™ 01-1111-41
74.20 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Alert:
Contains Sodium azide
Description
- Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
- This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
- Cross-reacts to some antibodies of rabbit origin, but have not been extensively tested for this reactivity
- Designed for use in compensation with all fluorochromes excited by blue (488nm), green (532nm), yellow-green (561nm), and red (633 to 635nm) lasers
- Compatible with eFluor™ 450 but is not optimized for compensation of other fluorochromes excited by a violet (405nm) laser
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Flow Cytometer | |
25 tests | |
Compensation Beads |
25 tests | |
eBeads™ |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2683425 | Certificate of Analysis | 29/01/2025 | 01-1111-41 |