Learn More
Hirschmann™ Opus™ Dispenser
Dispenses directly from the reagent bottle using a motor-powered reagent recirculation system. Hirschmann Instruments™ Opus™ Dispenser provides consistent results through electronic control.
Brand: Hirschmann™ 9581010
This item is not returnable.
View return policy
Description
- Electronic control and dispensing of volumes
- A light protection window for the valve block is also included in the scope of delivery
- TFT touchscreen with user-friendly menu
- Method memory for nine different dispensing procedures with storage of all settings
- Serial dispensing up to 9999 times
- Standard stepper function
- Media recirculating system for simple and rapid venting without reagent loss
- Free rotation
- Superior materials and reliable piston sealing, even in the case of problematic media
- Brown glass cylinder available for light-sensitive media
- USB and RS232 interface for PC connection
- Integration in automated procedures possible
Specifications
0.6% | |
10 mL | |
EU, US, UK plug type |
Electronic Bottletop Dispenser | |
TFT Touchscreen | |
100/240 V |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Product Content Correction
Product Title
Hirschmann™ Opus™ Dispenser >
Spot an opportunity for improvement?Share a Content Correction