Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Minimize the risks of improper measurement and contamination with these chemistry reagents, which are contained in self-filling vials.
Brand: Thermo Scientific™ AC4060
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors

Specifications
Silica | |
Orion AQUAfast AQ4000 Colorimeter | |
30 Auto-ID Ampoules | |
Colorimetric water and wastewater testing equipment | |
0.5 to 10 mg/L SiO2 | |
30 Ampoules |
Ampoule | |
AQUAfast silica Auto-ID ampoules, 30 pack | |
Heteropoly Blue | |
AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) | |
Drinking Water: ASTM D859-05,-10 / Standard Methods 4500-SiO2 D; Wastewater: ASTM D859-05 |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Aquafast IV silica
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.