Learn More
Saint Gobain Life Sciences Chemware™ Raschig Rings
Chemware™ Raschig Rings for use with distillation pumps in chemical processes and liquid or gas transfer applications.
Brand:  Saint Gobain Life Sciences D1069133
 View more versions of this product
Product Code. 10575434
  • £54.90 / Pack of 500
Explore more special offers
Description
Description
  • Packing for distillation columns and scrubbers.
  • Chemically inert except for molten alkali metals, fluorine, and strong fluorinating agents.
  • High surface area per unit of volume.
  • Minimum resistance to gas and liquid flow.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For chemical processes and liquid or gas transfer applications
Raschig Ring
For distillation columns
0.1 in.
3 mm
PTFE
3 mm
0.1 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Saint Gobain Life Sciences Chemware™ Raschig Rings > Chemware™ Raschig Rings; Dimensions (dia. x L): 3 x 3 mm