missing translation for 'onlineSavingsMsg'
Learn More
Osmium Tetroxide Solution, Honeywell™ Fluka™ Product Code.: 15688340

Osmium Tetroxide Solution, Honeywell™ Fluka™

Product Code. 15688340 Shop All Honeywell Fluka Products
2 mL, Glass Bottle
Click to view available options
Quantity:
2 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15688340

Brand: Honeywell-Fluka 2517552ML

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4 wt. % in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 20816-12-0
Quantity 2 mL
Molecular Formula O4Os
Linear Formula OsO4
MDL Number MFCD00011150
UN Number UN3287
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
SMILES O=[Os](=O)(=O)=O
IUPAC Name tetraoxoosmium
Molecular Weight (g/mol) 254.23
PubChem CID 30318
Formula Weight 254.23g/mol
Percent Purity 0%
Packaging Glass Bottle
Vapor Pressure 3.55psi (55°C), 1.23psi (20°C)
Density 1.0g/mL at 20°C, 1.04g/mL at 25°C
Chemical Name or Material Osmium Tetroxide Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Osmium Tetroxide Solution, Honeywell™ Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.