missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Baird-Parker Agar Base (Dehydrated)

Isolate and enumerate Staphylococcus aureus with this selective and sensitive medium.

Brand:  Thermo Scientific™ CM0275B

194.03 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10147992

  • £425.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Industrial Reference: AFNOR, AOAC, BSI, EP, IDF, ISO, NMKL, USDA.
Powder
500 g
Baird-Parker Agar Base
Agar
For 7.9L medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6245204 Certificate TypeCertificate of Analysis Date25/03/2025 Catalog NumberCM0275B
Lot Number6245203 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog NumberCM0275B
Lot Number6224626 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberCM0275B
Lot Number6224625 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberCM0275B
Lot Number6224391 Certificate TypeCertificate of Analysis Date22/02/2025 Catalog NumberCM0275B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Baird-Parker Agar Base (Dehydrated) > Baird-Parker Agar Base; 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only