Learn More
Thermo Scientific™ Lab-Lemco Nutrient Broth (Dehydrated)
Examine water, sewage, and dairy products by American standard methods using this nutritious medium.
Brand: Thermo Scientific™ CM0015B
106.73 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Examine water, sewage, and dairy products by American standard methods using Thermo Scientific™ Oxoid™ Lab-Lemaco Nutrient Broth (Dehydrated). It is used as a nutrient meat extract broth for general bacteriology.
Lab-Lemco Broth is a general purpose liquid medium used for the examination of water, dairy and food products1,2,3
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Specifications
Lab-Lemco Broth (Nutrient Agar) (US formulation) | |
Dehydrated Culture Media | |
For 62.5L medium |
Powder | |
500 g |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6211529 | Certificate of Analysis | 08/01/2025 | CM0015B |
3754578 | Certificate of Analysis | 07/12/2023 | CM0015B |
3652389 | Certificate of Analysis | 29/04/2023 | CM0015B |
3449584 | Certificate of Analysis | 05/04/2022 | CM0015B |
3289116 | Certificate of Analysis | 09/10/2021 | CM0015B |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Laboratory Use Only