Learn More
Eppendorf™ Centrifuge 5430/5430 R
The unique, cross-over Eppendorf Centrifuge 5430 and 5430 R combine the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, they can accommodate rotors for microplates and 15/50 mL conical tubes as well as Eppendorf tubes and PCR strips.
£3325.00 - £7360.00
Specifications
Display | Large, brightly lit LCD |
---|---|
Safety Features | Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition |
Timer | 30 sec. to 99:59 hrs., with continuous run function |
Certifications/Compliance | UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers |
Frequency | 50/60 Hz |
Product Code | Brand | Model | Refrigerated | Includes | Electrical Requirements | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Refrigerated | Includes | Electrical Requirements | Price | Quantity & Availability | ||
12883222
|
Eppendorf™
5427000267 |
Centrifuge 5430, keypad | No | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (GB) |
£3935.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12893222
|
Eppendorf™
5427000062 |
Centrifuge 5430, keypad | No | Without rotor | 230 V, 50/60 Hz (GB) |
£3485.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12853232
|
Eppendorf™
5427000461 |
Centrifuge 5430, rotary knobs | No | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (EU) |
£3935.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12843232
|
Eppendorf™
5427000666 |
Centrifuge 5430, rotary knobs | No | Without rotor | 230 V, 50/60 Hz (GB) |
£3485.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12803232
|
Eppendorf™
5428000066 |
Centrifuge 5430 R, keypad | Yes | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (GB) |
£7360.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12813232
|
Eppendorf™
5428000260 |
Centrifuge 5430 R, keypad | Yes | Without rotor | 230 V, 50/60 Hz (GB) |
£7090.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12823232
|
Eppendorf™
5428000465 |
Centrifuge 5430 R, rotary knobs | Yes | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (GB) |
£7360.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12833232
|
Eppendorf™
5428000660 |
Centrifuge 5430 R, rotary knobs | Yes | Without rotor | 230 V, 50/60 Hz (GB) |
£7090.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15835073
|
Eppendorf™
5428000060 |
Centrifuge 5430 R, keypad | Yes | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (GB) |
£7005.00
1 set |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15855073
|
Eppendorf™
5428000255 |
Centrifuge 5430 R, keypad | Yes | Without rotor | 230 V, 50/60 Hz (GB) |
£6755.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15875073
|
Eppendorf™
5428000460 |
Centrifuge 5430 R, rotary knobs | Yes | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (GB) |
£7005.00
1 set |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15895073
|
Eppendorf™
5428000655 |
Centrifuge 5430 R, rotary knobs | Yes | Without rotor | 230 V, 50/60 Hz (GB) |
£6755.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15875063
|
Eppendorf™
5427000260 |
Centrifuge 5430, keypad | No | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (GB) |
£3745.00
1 set |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15855063
|
Eppendorf™
5427000060 |
Centrifuge 5430, keypad | No | Without rotor | 230 V, 50/60 Hz (GB) |
£3325.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16602842
|
Eppendorf™
5427000186 |
Centrifuge 5430, rotary knobs | No | With Rotor A-2-MTP | 230 V, 50/60 Hz (GB) |
£4365.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15895063
|
Eppendorf™
5427000465 |
Centrifuge 5430, rotary knobs | No | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (GB) |
£3745.00
1 set |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15815073
|
Eppendorf™
5427000660 |
Centrifuge 5430, rotary knobs | No | Without rotor | 230 V, 50/60 Hz (GB) |
£3325.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Specifications
Large, brightly lit LCD | |
30 sec. to 99:59 hrs., with continuous run function | |
50/60 Hz |
Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition | |
UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers | |
230 V |