missing translation for 'onlineSavingsMsg'
Learn More
Palladium(II) nitrate, solution, Pd 10% w/w (cont. Pd), Thermo Scientific Chemicals Product Code.: 11480290

Palladium(II) nitrate, solution, Pd 10% w/w (cont. Pd), Thermo Scientific Chemicals

Product Code. 11480290
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11480290

Brand: Thermo Scientific Alfa Aesar 012621.NH

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Palladium(II) nitrate is used as a reactant for doping activated carbon for catalysis. It acts as a catalyst for hydrogenation of 1-heptyne and naphthalene. It serves as an important raw material for the preparation of platinum-palladium carbon alloy nanocatalysts for methanol-tolerant oxygen reduction reaction in fuel cells and in the synthesis of copper palladium alloy thin films. Furthermore, it is used in the synthesis of di-phenyl sulfide-modified palladium-titanium dioxide catalysts for acetylene hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-05-3
Molecular Formula N2O6Pd
Molecular Weight (g/mol) 230.43
MDL Number MFCD00011169
InChI Key GPNDARIEYHPYAY-UHFFFAOYSA-N
Synonym palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrateShow MoreShow Less
PubChem CID 24932
IUPAC Name palladium(2+);dinitrate
SMILES [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Brown to Orange
UN Number UN3264
Concentration Pd 10% w/w (cont. Pd)
Solubility Information Miscible with water and dilute nitric acid.
Formula Weight 230.43
Quantity 1 g
Physical Form Liquid
Chemical Name or Material Palladium(II) nitrate, Solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06J049 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number012621.ND,012621.NH,012621.NB
Lot NumberP24J054 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number012621.NH
Lot NumberW20E022 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012627.NA,012621.ND,012621.NH,012621.NB
Lot NumberU07B071 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012627.NA,012621.ND,012621.NH,012621.NB
Lot NumberE02R001 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012627.NA,012621.ND,012621.NH,012621.NB

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Palladium(II) nitrate, solution, Pd 10% w/w (cont. Pd), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.