missing translation for 'onlineSavingsMsg'
Learn More
Peri-xanthenoxanthene, 95%, Thermo Scientific™ Product Code.: 15294408

Peri-xanthenoxanthene, 95%, Thermo Scientific™

Product Code. 15294408
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15294408

Brand: Thermo Scientific Acros 454892500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 191-28-6
Molecular Formula C20H10O2
Molecular Weight (g/mol) 282.3
InChI Key AMDQVKPUZIXQFC-UHFFFAOYSA-N
Synonym dinaphthylene dioxide, peri-xanthenoxanthene, dinaphthalene dioxide, xantheno 2,1,9,8-klmna xanthene
PubChem CID 240798
SMILES C1=CC2=C3C(=C1)OC4=C5C3=C(C=C2)OC6=CC=CC(=C65)C=C4

Specifications

Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 282.3
Percent Purity 95%
Chemical Name or Material Peri-xanthenoxanthene
compliance-icons
Product Identifier
  • Peri-xanthenoxanthene, 95%
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Peri-xanthenoxanthene, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.