Learn More
pH Buffers, Twin Neck, Fisherbrand™
Perform accurate pH calibrations with color-coded, twin neck Fisherbrand™ pH Buffer Bottles.
Brand: Fisherbrand BS10/500ML
11.95 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Perform accurate pH calibrations with color-coded, twin neck Fisherbrand™ pH Buffer Bottles.
Buffer Color Codes:
- Pink = pH 4.01
- Yellow = pH 7.00
- Blue = pH 10.01
Available in 500 mL bottles

Specifications
pH calibration | |
10.01 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.