Learn More
Christeyns™ Phago'Solution
Phago'Solution is intended for hygienic and surgical hand disinfection as an alternative to simple or hygienic hand washing in the absence of a water point, during serial contact, and before any aseptic gesture.
Brand: Christeyns™ C65045
View more versions of this product
This item is not returnable.
View return policy
Description
- Antimicrobial activity in 30 sec.
- Dermatologically tested
- Promotes a high level of compliance
Specifications
Hand Disinfectant | |
Liquid | |
1 L |
Colorless | |
12 Airless Bottles |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
SDS_17870255_EN.pdf
File size: 167.4 kB
Product Content Correction
Product Title
Christeyns™ Phago'Solution > Phago'Solution; Volume (Metric): 1 L
Spot an opportunity for improvement?Share a Content Correction