missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ EDC, No-Weigh™ Format

Immobilize or conjugate proteins using 1-Ethyl-3-(3-dimethylaminopropyl) carbodiimide (CAS 25952-53-8), a water-soluble, carboxyl-to-amine crosslinker.

Brand:  Thermo Scientific™ A35391


 View more versions of this product

Product Code. 15781819

  • £159.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce EDC is a water-soluble carbodiimide crosslinker that activates carboxyl groups for spontaneous reaction with primary amines, enabling peptide immobilization and hapten-carrier protein conjugation.

Thermo Scientific No-Weigh products are specialty reagents provided in pre-aliquoted format. The pre-weighed packaging helps prevent loss of reagent reactivity and contamination over time by minimizing repetitive opening and closing of vial. The format enables use of fresh vial of reagent each time, eliminating hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

  • Reactive group: Carbodiimide
  • Reaction target: Activates carboxyl groups to conjugate to amino groups (primary amines)
  • Several conjugation strategies: React EDC alone with target groups or include NHS or Sulfo-NHS to increase reaction efficiency or to stabilize active intermediate for later reaction to amines
  • Neutral linkage: Forms neutral amide bonds between carboxyls and amines
  • Water-soluble Reagent: Add directly to reactions in aqueous, physiological buffers
  • Soluble reaction byproducts: Easily removed by washing with water or dilute acid
  • High-purity, crystalline reagent: Use to create high-quality, activated derivatives
Applications:
  • Conjugate carboxyl and amino groups among peptides and proteins
  • Couple haptens to immunogenic carrier proteins (e.g., attach a peptide to KLH)
  • Immobilize peptide antigens to affinity-purify antibodies
  • Create NHS-activated, amine-reactive labeling compounds
  • Crosslink proteins to carboxyl coated beads or surfaces
  • Activate nanoparticles with amine-reactive Sulfo-NHS esters
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amine-Carboxyl
EDC
No
Upon receipt store desiccated at -20°C.
Wet Ice
Chemical Labeling
Carbodiimide
Powder
Water
Yes
No
191.7
0.0 Å
No
No-Weigh™, Pierce™
Heterobifunctional
Short (<10 Å)
10 x 1 mg
Standard, Single-use, Premium-grade
Crosslinker
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL402614 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA35391
Lot NumberZL403561 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog NumberA35391
Lot NumberZJ401296 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA35391
Lot NumberZK401297 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA35391
Lot NumberZK401297A Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA35391
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    59

    Hours

    2

    Minutes

    27

    Seconds

    21
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ EDC, No-Weigh™ Format > 10 x 1 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.