Learn More
Thermo Scientific™ Immobilized Trypsin, TPCK Treated (Agarose Resin)
Cleave the carboxyl-side of Arg and Lys residues with trypsin that isTPCK-treated to block chymotrypsin activity; its immobilized form allows enzyme removal after digestion.
Brand: Thermo Scientific™ 20230
110.20 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ Pierce TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.
Highlights:
- Immobilized TPCK Trypsin eliminates trypsin contamination in digests
- TPCK inhibits chymotrypsin activity

Specifications
Pierce Immobilized Trypsin, TPCK Treated | |
Liquid Suspension | |
Trypsin |
Contents: Immobilized TPCK Trypsin, 2 mL of settled gel supplied as a 50% slurry containing glycerol and 0.05% sodium azide. Storage: Upon receipt store at 4°C. Product is shipped at ambient temperature. |
|
2 mL |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
38Hours
11Minutes
4Seconds
48New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.