Learn More
Eppendorf™ Pipette Holder 2
GREENER_CHOICE
Eppendorf™ Pipette Holder 2 keeps your Eppendorf™ pipette within good reach and saves precious bench space.
Brand:  Eppendorf™ 3116000112
30.39 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Product Code. 15789348
  • £32.50 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Eppendorf™ Pipette Holder 2 keeps your Eppendorf™ pipette within good reach and saves precious bench space.
  • Carries one manual single or multi-channel pipette including Eppendorf Research™, Research™ plus, Reference™, Reference™ 2 or Biomaster™
  • Designed for wall mounting or for installation in Pipette Carousel 2 and Charger Carousel 2
  • Sticky tape for wall mounting included
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sticky tape for wall mounting
For wall mounting or for installation in Pipette Carousel 2 and Charger Carousel 2
One manual single or multi-channel pipette including Eppendorf Research™, Research™ plus, Reference™, Reference™2 or Biomaster™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Pipette Holder 2 > For use with: One manual single or multi-channel p