Learn More
Fisherbrand™ Polystyrene Semi Micro Cuvettes for UV and Visible Wavelengths
Description
- Very low variation of extinction values
- Excellent optical transmission range
- Cavity-sorted production
Specifications
Semi-micro Cuvette | |
1.6 mL | |
100/Pk. | |
45 mm |
Reusable | |
Plastic | |
220 to 900 nm | |
12.5 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Product Title
Fisherbrand™ Polystyrene Semi Micro Cuvettes for UV and Visible Wavelengths > Capacity: 1.6mL; Wavelength: 220 to 900nm; QTY: 100/Pk.
Spot an opportunity for improvement?Share a Content Correction