Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, SYBR Safe 2%
Designed for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel SYBR Safe 2% agarose gels.
Brand: Invitrogen™ G8152ST
Includes: E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
Description
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, SYBR Safe 2%, provides everything you need to quickly start using precast 2% E-Gel agarose gels with SYBR Safe on the device.
The kit includes:
- 18 E-Gel agarose gels with SYBR Safe, 2% (Cat. No. G521802)
- E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
- E-Gel 50 bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
Services
1 year warranty

Specifications
• 1 electrophoresis device • 18 E-Gel SYBR Safe 2% gels • 1 vial E-Gel 50 bp DNA Ladder • 1 power adapter • 1 pair of viewing glasses • 1 E-Gel Go! Adapter Store at room temperature. |
|
Electrophoresis Device Starter Kit | |
240 V AC (Power Supply), 48 V DC (Electrophoresis Device) |
Mini | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Invitrogen E-Gel Power Snap electrophoresis system features
-
Invitrogen Power Snap camera features
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.