missing translation for 'onlineSavingsMsg'
Learn More
(±)-Propylene oxide, 99.5%, extra pure, Thermo Scientific Chemicals Product Code.: 10771335

(±)-Propylene oxide, 99.5%, extra pure, Thermo Scientific Chemicals

Product Code. 10771335
2.5 L, Glass bottle
Click to view available options
Quantity:
5 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10771335

Brand: Thermo Scientific Acros 220160025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-56-9
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00005126
InChI Key GOOHAUXETOMSMM-UHFFFAOYNA-N
Synonym propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropaneShow MoreShow Less
PubChem CID 6378
ChEBI CHEBI:38685
IUPAC Name 2-methyloxirane
SMILES CC1CO1

Specifications

Melting Point -112.0°C
Density 0.8300g/mL
Boiling Point 34.0°C
Flash Point −37°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Refractive Index 1.3650 to 1.3670
Quantity 2.5 L
UN Number 1280
Beilstein 17, 6
Merck Index 15, 7969
Specific Gravity 0.83
Solubility Information Solubility in water: 40g/100mL (20°C). Other solubilities: miscible with alcohol and ether
Viscosity 0.32 mPa.s (20°C)
Formula Weight 58.08
Percent Purity 99.5%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material (±)-Propylene oxide, Extra Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467436 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number220160050,220160010,220160025
Lot NumberA0467435 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number220160050,220160010,220160025
Lot NumberA0464378 Certificate TypeCertificate of Analysis Date04/07/2024 Catalog Number220160050,220160010,220160025
Lot NumberA0461061 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number220160050,220160010,220160025
Lot NumberA0439239 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number220160050,220160010,220160025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (+/-)-Propylene oxide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(±)-Propylene oxide, 99.5%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.