Learn More
Cole-Parmer™ PTFE Screen Gaskets
Protect your pump, valves and other components in fluid line
Brand: Cole-Parmer™ 3053886
This item is not returnable.
View return policy
Description
- Pre-filter before critical membrane filters
- Generate turbulence for enhanced mass or heat transfer
- Purify in chromatography applications
Specifications
100 | |
3.81 cm | |
PTFE | |
PTFE Gasket |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Product Title
Cole-Parmer™ PTFE Screen Gaskets > Cole-Parmer Screen Gasket; Mesh size: 80;Particle size retained: 140 microns;Sanitary gasket size: 1.5 in.
Spot an opportunity for improvement?Share a Content Correction