Learn More
Fisherbrand™ PTFE Screw Propeller Stirrer Shaft
Use with shaft guides. The PTFE anchor stir shafts have a stainless-steel core.
£48.05 - £222.86
Specifications
For Use With (Equipment) | Universal Stirrer Guide |
---|---|
For Use With (Application) | Mixing |
Quantity | 1 Each |
Temperature (Metric) | -200°C to +280°C |
Product Code | Brand | Diameter (Metric) Shaft | Length (Metric) | Diameter (Metric) Fan | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Shaft | Length (Metric) | Diameter (Metric) Fan | Price | Quantity & Availability | ||||
10779372
|
Fisherbrand™
005.230.8.2 |
8 mm | 300 mm | 40 mm |
£70.10
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
10609352
|
Fisherbrand™
005.265.2 |
9.5 mm | 650 mm | 60 mm |
£77.15
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||
10689152
|
Fisherbrand™
005.240.8.2 |
8 mm | 400 mm | 40 mm |
£106.00
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10719762
|
Fisherbrand™
005.250.8.2 |
8 mm | 500 mm | 50 mm |
£85.45
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||
10688962
|
Fisherbrand™
005.240.2 |
6 mm | 400 mm | 40 mm |
£48.05
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||
10740143
|
Fisherbrand™
005.255.2 |
9.5 mm | 550 mm | 60 mm |
£83.65
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||
10075991
|
Fisherbrand™
005.250.2 |
6 mm | 500 mm | 50 mm |
£60.45
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
10127991
|
Fisherbrand™
005.230.2 |
6 mm | 300 mm | 40 mm |
£56.65
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
10279841
|
Fisherbrand™
005.255.10.2 |
10 mm | 550 mm | 70 mm |
£186.16
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11563252
|
Fisherbrand™
005.02100.16.2 |
16 mm | 1000 mm (EX) | 100 mm |
£222.86
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||
10701493
|
Fisherbrand™
005.0265.10.2 |
10 mm | 650 mm (EX) | 70 mm |
£112.00
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
10478401
|
Fisherbrand™
005.0275.2 |
9.5 mm | 750 mm (EX) | 60 mm |
£79.70
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||
10711683
|
Fisherbrand™
005.0275.10.2 |
10 mm | 750 mm (EX) | 70 mm |
£94.30
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||
10620503
|
Fisherbrand™
005.02100.12.2 |
12 mm | 1000 mm (EX) | 80 mm |
£135.00
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||
10585682
|
Fisherbrand™
005.0275.12.2 |
12 mm | 750 mm (EX) | 80 mm |
£103.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||
10351542
|
Fisherbrand™
005.0265.2 |
9.5 mm | 650 mm (EX) | 60 mm |
£158.00
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
10762603
|
Fisherbrand™
005.0265.12.2 |
12 mm | 650 mm (EX) | 80 mm |
£114.40
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
Description
PTFE shaft stirrers have a unique, fully molded construction featuring a rotor molded to a fully PTFE-encapsulated steel reinforced drive shaft.- Chemically resistant, inert, and leak-free
- Easy to clean
- No cross contamination
- Shatterproof
- Non-scratch
- Use in temperatures up to approximately 280°C
- Exposed shaft models (EX) are produced with a stainless steel core with an exposed end for more rigid clamping to the chuck drive
Specifications
Universal Stirrer Guide | |
Mixing | |
1 Each | |
-200°C to +280°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ PTFE Screw Propeller Stirrer Shaft