missing translation for 'onlineSavingsMsg'
Learn More
Pyrazolo[1,5-a]pyridine-7-carboxaldehyde, 97%, Thermo Scientific Chemicals Product Code.: 11474480

Pyrazolo[1,5-a]pyridine-7-carboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11474480
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11474480

Brand: Thermo Scientific Alfa Aesar H50074.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 362661-83-4
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD06739029
InChI Key SGNBVQKSJWCGOJ-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyridine-7-carbaldehyde, pyrazolo 1,5-a pyridine-7-carboxaldehyde, 8-hydropyrazolo 1,5-a pyridine-7-carbaldehyde, pyrazolo 1,5-a pyridine-7-carboxaldehyde 9ci, pyrazolo 1,5-alpha pyridine-7-carboxaldehydeShow MoreShow Less
PubChem CID 10866474
IUPAC Name pyrazolo[1,5-a]pyridine-7-carbaldehyde
SMILES C1=CC2=CC=NN2C(=C1)C=O

Specifications

Melting Point 83°C to 86°C
Quantity 250 mg
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material Pyrazolo[1,5-a]pyridine-7-carboxaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD11R011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50074.03,H50074.MD
Lot NumberQ12C005 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50074.03,H50074.MD
Lot Number10161588 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50074.03,H50074.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrazolo[1,5-a]pyridine-7-carboxaldehyde, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.