Learn More
Pyrex™ Gas Distribution Tube
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
Brand:  Pyrex 3830/00M
79.96 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10217634
  • £157.00 / Pack of 2
Estimated Shipment: 13-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
  • Ideal for the dispersion of gas in liquids
  • Borosilicate glass tube with sintered thimble
  • Choice of thimble porosities to achieve optimum gas bubble size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Porosity: 0
300 mm
Sintered thimble
Borosilicate Glass (3.3)
For the dispersion of gas in liquids
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pyrex™ Gas Distribution Tube > Outer Thimble Diameter: 15mm; Length: 300mm; Pore Size: 160 to 250μ ; Outer Stem Diameter: 8mm