missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ GFP (eGFP) siRNA

Silencer™ GFP (eGFP) siRNA

Brand:  Invitrogen™ AM4626

138.00 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


134.67 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10025764

  • £207.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Invitrogen™ Silencer™ GFP (eGFP) Positive Control siRNA is ideal for developing and optimizing siRNA experiments. 5 nmol is provided, along with an additional tube of 2 nmol negative control siRNA.
  • Validated siRNA controls for optimizing siRNA experiments
  • Gene-specific control siRNAs provided with scrambled, negative controls
  • Functionally tested in several common cell lines
  • HPLC purified, duplexed, and ready to use Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Invitrogen™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Store at or below -20°C. Do not store in a frost-free freezer.
    siRNA
    Unconjugated
    siRNA
    5 nmol, 2 nmol
    GFP (Green Fluorescent Protein)
    Positive Control
    Transfection, RNAi
    Silencer™, Ambion™
    HPLC
    Dry Ice
    Suggerimenti di prodotto

    Suggerimenti di prodotto

    Videos
    SDS
    Documenti

    Documenti

    Certificati

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    2 results found
    Lot Number Certificate Type Date Catalog Number
    Lot NumberASO0WJTA Certificate TypeCertificate of Analysis Date15/04/2017 Catalog NumberAM4626
    Lot NumberASO0RP8F Certificate TypeCertificate of Analysis Date01/04/2017 Catalog NumberAM4626
    Offerte speciali

    Offerte speciali

    Correzione del contenuto del prodotto

    Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

    Titolo del prodotto
    Invitrogen™ Silencer™ GFP (eGFP) siRNA > 5 nmol

    Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

    Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato

    For Research Use Only. Not for use in diagnostic procedures.