Learn More
Thermo Scientific™ QuEChERS AOAC 2007.01 Method Extraction Kits
Use Thermo Scientific™ QuEChERS AOAC 2007.01 Method Extraction Kits to prepare complex samples for LC-MS or GC-MS analysis of residues, toxins, and other contaminants.
Brand: Thermo Scientific™ S1-15-AOAC-POT
117.80 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the AOAC 2007.01 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.
QuEChERS AOAC 2007.01 Method Extraction Kits
Extract analytes of interest from a homogenized sample using extraction salts. Use these kits with 10 g or 15 g samples. Choose from:
- Extraction Salts in Pouches, No Tubes
- Extraction Salts in Pouches with 50 mL Tubes
- Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers
Chemicals used in the AOAC 2007.01 Method
- Acetonitrile: used to perform single-phase extraction on a sample
- Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning; removes remaining water from sample during dSPE
- NaOAc: buffers the sample to stabilize pH
- PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids from sample during dispersive solid phase extraction
- C18: removes fats, sterols, and other non-polar interferences from sample
- GCB: graphitized carbon black, removes pigment (not recommended for use with planar pesticides)

Specifications
QuEChERS AOAC Extraction Kit for 15 g sample; 6 g MgSO4, 1.5 g NaOAC in Pouches; No Tubes; 50/Pk. | |
Salts in Pouches | |
6 g MgSO4, 1.5 g NaOAC | |
None |
QuEChERS Extraction Kit | |
QuEChERS sample preparation for AOAC 2007.01 Method | |
15 g sample |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
26Hours
14Minutes
45Seconds
23 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
57Hours
14Minutes
45Seconds
23New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.