Learn More
Gilac™ Rectangular HACCP Container with Lid
Rectangular HACCP Container with Lid is a food container for large storage volumes; lid can support the weight of heavy loads.
Brand: Gilac™ G119809
This item is not returnable.
View return policy
Description
- HACCP rectangular food container.
- For large storage volumes, lid can support the weight of heavy loads.
- Multipurpose container.
- Rewritable identification cartridge for inventory tracking.
- No gaps nor overlaps.
- Machine washable.
- Recessed handles for better handling.
- Stackable without lid.
- Compatible with all GILAC 4 and 6 wheel dollies.
Specifications
Storage Tote | |
Red | |
9.2 gal. |
HDPE | |
Rectangular | |
35 L |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Gilac™ Rectangular HACCP Container with Lid > Rectangular HACCP Container with Lid; Capacity: 35 L; Color: Red
Spot an opportunity for improvement?Share a Content Correction