missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PathoDX™ Strep B Grouping Latex

PathoDx Strep B Grouping Latex is used with Strep Grouping Kits.

Brand:  Thermo Scientific™ R62031

91.52 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 12613728

  • £94.80 / 60 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ Remel™ PathoDx™ and Strep B Grouping Latex with Strep Grouping Kits. The PathoDx procedure utilizes a latex agglutination method in conjunction with a nitrous acid extraction procedure1,2. The IgG coupled to the latex is highly specific for a given streptococcal group antigen. This method offers significant advantages over other streptococcal grouping procedures in terms of rapidity, simplicity, and convenience.

The streptococcal group carbohydrates of streptococcal group B is a complex antigen with a rhamnose oligosaccharide and differing side chains, consisting primarily of glucosamine, either acetylated or non-acetylated. In the PathoDx procedure, specific antibody on latex particles reacts with, and agglutinates, streptococcal group antigen extracted from the bacterial cell wall. The test is intended for use with streptococcal colonies that are betahemolytic on sheep blood agar or for use with pure streptococcal isolates in broth cultures. The reagents are designed to give a positive (2+ or greater) agglutination with one to two 18 to 24 hour colonies for most beta-hemolytic streptococcal isolates.

  • Ready-to-use – Prepared reagents
  • Easy-to-store – Store at 2-8°C
  • Easy to interpret – The PathoDx Strep Grouping kit is designed to give a 2+ or greater agglutination reaction with the extract of one or two colonies of an 18 to 24 hour culture of streptococci of Lancefield group B (large colony variety) in 60 seconds for most streptococcal isolates. In a negative test, a uniform milky appearance with no agglutination after 60 seconds.

Strep B Grouping Latex (Pink Cap) contains 3.0mL of a suspension of synthetic blue latex particles coated with rabbit antibody IgG-sensitized with 0.098% sodium azide and 0.05% ProClin 300® as preservatives.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PathoDX™ Strep B Grouping Latex
Streptococcus Test Kit
60 Tests/Pk.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

SDS
Documents

Documents

Instructions

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6211154 Certificate TypeCertificate of Analysis Date15/04/2025 Catalog NumberR62031
Lot Number6194554 Certificate TypeCertificate of Analysis Date08/03/2025 Catalog NumberR62031
Lot Number6194553 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberR62031
Lot Number6171048 Certificate TypeCertificate of Analysis Date09/01/2025 Catalog NumberR62031
Lot Number6159420 Certificate TypeCertificate of Analysis Date12/12/2024 Catalog NumberR62031
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ PathoDX™ Strep B Grouping Latex > Strep B Grouping Latex

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.