Learn More
Thermo Scientific™ Rotor Packages for Sorvall™ ST8 Benchtop Centrifuge
Convenient cell culture and clinical packages
Brand: Thermo Scientific™ 75005761
2465.25 GBP valid until 2025-12-31
Use promo code "24378" to get your promotional price.
Description
Convenient cell culture and clinical packages
Rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange to certified Thermo Scientific™ ClickSeal™ biocontainment lids—shorten run set-up time while providing peace-of-mind that your samples are secure.
Secure, push-button Auto-Lock rotor exchange in less than 3 seconds delivers:
- Trouble-free rotor installation and removal:
- No tools are required
- No complicated rotor changing procedures
- Clear chamber access for cleaning convenience
- Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
- Biocontainment sealing, including certified ClickSeal lids for glove-friendly, one-handed operation
- 8 adapters available to meet your evolving clinical and research needs

Specifications
Aluminum | |
8 x 50 mL | |
4,500 rpm | |
12.4 cm | |
Thermo Scientific™ Sorvall™ ST8 Benchtop Centrifuge | |
TX-150 Rotor (75005701), Set of 4 Conical Buckets (75005703), Set of 2 Adapters for 15mL Conical Tubes (75005808) |
Swinging Bucket Rotor | |
3,260 x g | |
5.1 to 14.4 cm | |
1.8 cm | |
90° | |
Yes |
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.