Learn More
SCAT™ Safety Waste Caps
Safety Waste Caps for the safe disposal of liquid waste. For maximum protection and efficient work with hazardous waste liquids. Offers multiple options to connect different tubing and capillary sizes.
£104.00 - £223.00
Product Code | Brand | Size | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Includes | Price | Quantity & Availability | |||||
10681714
![]() |
SCAT™
107037 |
B53 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
£160.00
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
10529850
![]() |
SCAT™
107038 |
B63 | 1 x 2.3/3.2 mm OD, 3 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
£207.00
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
|||||
10880191
![]() |
SCAT™
107051 |
B63 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
£123.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10170510
![]() |
SCAT™
107050 |
B63 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
£123.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10785821
![]() |
SCAT™
107036 |
B83 | 4 x 2.3/3.2 mm OD Connector for Exhaust Filter |
£212.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10619993
![]() |
SCAT™
107034 |
B83 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
£223.00
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
|||||
10119164
![]() |
SCAT™
108025 |
S50 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
£144.00
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
10614237
![]() |
SCAT™
108023 |
S50 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
£128.00
Each |
In Stock
Log in to see stock. |
|||||
10377609
![]() |
SCAT™
108024 |
S50 | 3 x 2.3/3.2 mm OD, Grounding Connection, Connector for Exhaust Filter |
£178.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10272723
![]() |
SCAT™
107922 |
S51 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
£142.00
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
10265103
![]() |
SCAT™
107930 |
S51 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
£123.00
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
10605645
![]() |
SCAT™
107924 |
S55 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
£177.00
Each |
In Stock
Log in to see stock. |
|||||
10699515
![]() |
SCAT™
107917 |
S55 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
£123.00
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
10717595
![]() |
SCAT™
108047 |
S65 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
£156.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10218784
![]() |
SCAT™
108046 |
S65 | 5 x 2.3/3.2 mm OD Connector for Exhaust Filter |
£160.00
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
|||||
10629993
![]() |
SCAT™
107947 |
S90 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
£203.00
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
10508625
![]() |
SCAT™
107927 |
S90 | 4 x 2.3/3.2 mm OD Connector for Exhaust Filter |
£145.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
12655895
![]() |
SCAT™
107257 |
S95 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
£200.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
10580241
![]() |
SCAT™
107926 |
S70/71 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
£123.00
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
|||||
10861511
![]() |
SCAT™
107913 |
S70/71 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
£123.00
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
17842115
![]() |
SCAT™
108143 |
S55 | 1 x 2.3/3.2 mm OD, 3 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
£104.00
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
17927250
|
SCAT™
108142 |
£167.00
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||||
17947260
|
SCAT™
108177 |
£156.00
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||||
Description
- 360° free turning PTFE body, resistant against aggressive media
- Exhaust filter system against harmful vapors
- Fittings and tube connectors in different sizes
- Flexible system
- Time saving in changing the waste containers while tubes are fixed
- Lab safety, technical protective measures, optimal protection when collecting liquid waste
- Cost efficiency
- Huge range of accessories, optional: funnel use, level control preventing overflowing containers
- Huge portfolio of thread sizes suitable for waste containers from different manufacturers
- Safety disposal of HPLC waste
- High work performance
- Personal protection, avoid skin contact with organic solvents, occupational safety, ergonomics
- Sealed bottles and containers
- Create a comfortable working climate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
SCAT™ Safety Waste Caps